Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAME hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
1784-03-8
C14H23ClN4O4S
378.872
JIQFFACVQXXHMY-YDALLXLXSA-N
tos-arg-ome.hcl,tame hydrochloride,tos-arg-ome hcl,nalpha-tosyl-l-arginine methyl ester hydrochloride,nalpha-p-tosyl-l-arginine methyl ester hydrochloride,nalpha-p-toluenesulfonyl-l-arginine methyl ester hydrochloride,tos-arg-omehcl,s-methyl 5-guanidino-2-4-methylphenylsulfonamido pentanoate hydrochloride,tosyl-l-arginine methyl ester hydrochloride,n-4-tosyl-l-arginine methyl ester hydrochloride
2723792
methyl (2S)-5-(diaminomethylideneamino)-2-[(4-methylphenyl)sulfonylamino]pentanoate;hydrochloride
CC1=CC=C(C=C1)S(=O)(=O)NC(CCCN=C(N)N)C(=O)OC.Cl
This item is not returnable. View return policy
1784-03-8
C14H23ClN4O4S
378.872
JIQFFACVQXXHMY-YDALLXLXSA-N
tos-arg-ome.hcl,tame hydrochloride,tos-arg-ome hcl,nalpha-tosyl-l-arginine methyl ester hydrochloride,nalpha-p-tosyl-l-arginine methyl ester hydrochloride,nalpha-p-toluenesulfonyl-l-arginine methyl ester hydrochloride,tos-arg-omehcl,s-methyl 5-guanidino-2-4-methylphenylsulfonamido pentanoate hydrochloride,tosyl-l-arginine methyl ester hydrochloride,n-4-tosyl-l-arginine methyl ester hydrochloride
2723792
methyl (2S)-5-(diaminomethylideneamino)-2-[(4-methylphenyl)sulfonylamino]pentanoate;hydrochloride
CC1=CC=C(C=C1)S(=O)(=O)NC(CCCN=C(N)N)C(=O)OC.Cl

Ubiquitin ligase APC/C inhibitor; promotes Cdc20 autoubiquitination

Competitive inhibitor of the ubiquitin ligase anaphase-promoting complex/cyclosome (APC/C); promotes Cdc20 autoubiquitination. Inhibits cyclin proteolysis in mitotic Xenopus oocyte extracts (IC50 = 12 μM) and blocks cyclin degradation in interphase extract.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 378.87
Percent Purity >99%
Chemical Name or Material TAME hydrochloride

Recommended Storage : Store at -20°C