Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tamoxifen Citrate, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
1 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
54965-24-1
C32H37NO8
563.65
FQZYTYWMLGAPFJ-OQKDUQJOSA-N
(2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid
OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1
54965-24-1
C32H37NO8
563.65
FQZYTYWMLGAPFJ-OQKDUQJOSA-N
(2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid
OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1

T1423, 54965-24-1, C26H29NO•C6H8O7

Spectrum™ Tamoxifen Citrate, USP is a chemotherapeutic agent used to treat breast cancer. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.002
Loss on Drying 0.005
Quantity 1 kg
CAS Max % 1
Percent Purity 99 to 101%
Grade USP