Learn More
Fisherbrand™ Tanks with Dipping Baskets
Perforated bottom and sides to fit in tanks.
$203.00 - $259.00
Specifications
Material Polyethylene
Includes Fabricated polypropylene dipping basket and cover
Type Tank
View More Specs
Products 3
Catalog Number Capacity (English) Diameter (English) Outer Height (English) Width (English) Price Quantity & Availability  
Catalog Number Capacity (English) Diameter (English) Outer Height (English) Width (English) Price Quantity & Availability  
S37129 Products for Science Education View Documents 6 gal. 13 in. 16 in. 13 in. Each for $203.00
Add to cart
S37130 Products for Science Education View Documents 10 gal. 15.5 in. 18 in. 15.2 in. Each for $259.00
Add to cart
S37127 Products for Science Education View Documents 3 gal. 11.5 in. 10.8 in. 11.5 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Dipping baskets with 3 to 11 gallon capacity tanks
  • Low density polyethylene tanks
  • With covers and specially fabricated polypropylene dipping baskets
  • Dipping baskets have strap handle
Specifications
Specifications
Polyethylene
Fabricated polypropylene dipping basket and cover
Tank