Learn More
Tankyrase 1 Chemiluminescent Activity Assay, R&D Systems
a high-throughput, semi-quantitative ELISA designed to measure Tankyrase 1 activity
Manufacturer:  R&D Systems 4701192K
 View more versions of this product
Catalog No. 4701192K
  Check Availability
Add to cart
Includes:
  • Histone-coated 96-stripwell Microplate
  • Anti-PAR Monoclonal Antibody
  • HRP-conjugated Goat Anti-Mouse Antibody
  • Tankyrase 1
  • XAV939a
  • Diluents & Buffers
  • Unmodified NAD Substrate
  • HRP Substrat
Specifications
Description
Features
  • Amenable to high-throughput
  • Highly sensitive: capable of measuring 0.1 mUnits of Tankyrase 1
  • 3 hour assay time
  • Provides full length Tankyrase 1 enzyme and unmodified NAD substrate
Description & Specifications
Specifications
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA kit
0.1mU
192 Tests
ELISA
1 kit
Multi-species