Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Tankyrase 1 Colorimetric Activity Assay
a high-throughput, semi-quantitative ELISA designed to measure Tankyrase 1 activity.
Supplier: R&D Systems™ 4700096K
Includes:
- Histone-coated 96-stripwell Microplate
- Anti-PAR Monoclonal Antibody
- HRP-conjugated Goat Anti-Mouse Antibody
- Tankyrase 1
- XAV939a
- Diluents & Buffers
- Unmodified NAD Substrate
- HRP Substrat
Description
Tankyrase 1, also known as PARP5a, is a Poly (ADP-ribose) Polymerase (PARP) that associates with telomeres via its interaction with the telomeric-repeat binding factor TRF1. This assay utilizes a histone-coated 96-well microplate and an anti-poly (ADP-rib
Features- Amenable to high-throughput
- Highly sensitive: capable of measuring 0.1 mUnits of Tankyrase 1
- 3 hour assay time
- Provides full length Tankyrase 1 enzyme and unmodified NAD substrate
Specifications
0.1mU | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
96 Tests | |
ELISA |
HRP | |
ELISA Kit | |
Multi-species | |
1 Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction