Learn More
Tankyrase Polyclonal Antibody, Invitrogen™
Description
PA1-32225 detects Tankyrase from human samples. PA1-32225 has been successfully used in Western blot applications. The PA1-32225 immunogen is: Synthetic peptide internal to tankyrase.
Specifications
Specifications
| Antigen | Tankyrase |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.1 mg/mL |
| Conjugate | Unconjugated |
| Gene | TNKS |
| Gene Accession No. | O95271 |
| Gene Alias | PARP-5a, PARP5A, PARPL, TIN1, TINF1, TNKS1, pART5 |
| Gene Symbols | TNKS |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.