Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tannic Acid, Reagents
Click to view available options
Quantity:
100 g
500 g
Specifications
Specifications
Color | Off-White to Light Brown |
Grade | Reagent |
Quantity | 500 g |
Packaging | Poly Bottle |
Physical Form | Solid |
Specific Gravity | 2.807 |
Chemical Name or Material | Tannic Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction