Learn More
Tanshinone IIA, Tocris™
Inhibits NF-κB and AP-1 DNA binding; Displays antioxidant properties
Manufacturer:  R&D Systems 4426/50
Quantity 50mg
 View more versions of this product
Catalog No. 44-265-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
294.34
Tanshinone IIA
C19H18O3
6,7,8,9-Tetrahydro-1,6,6-trimethylphenanthro[1,2-b]furan-10,11-dione
Specifications
568-72-9
C19H18O3
6,7,8,9-Tetrahydro-1,6,6-trimethylphenanthro[1,2-b]furan-10,11-dione
Tanshinone IIA
>98%
294.34
50mg
Safety and Handling
Recommended Storage : Store at Room Temperature