Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tantalum carbide, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 1214418

Quantity 50g

 View more versions of this product

Catalog No. AA1214418


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12070-06-3
MFCD00011255
Tantalum carbide
192.96
UN3178

Specifications

12070-06-3
192.96
UN3178
−325 Mesh
5500°C
3880°C
Tantalum carbide
Powder
MFCD00011255
13.9g/mL
Odorless
Gray to black
50g
99.5%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit