Alfa Aesar™ Tantalum gauze, 100 mesh woven from 0.08mm (0.003 in.) dia. wire, twilled weave

Manufacturer: Alfa Aesar™ 46695GT

Quantity 150x150mm

 View more versions of this product

Catalog No. AA46695GT

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-25-7
    MDL Number MFCD00011252
    Chemical Name or Material Tantalum gauze, 100 mesh woven from 0.08mm (0.003 in.) dia. wire, twilled weave

    Specifications

    CAS 7440-25-7
    TSCA TSCA
    Physical Form Wire Cloth
    MDL Number MFCD00011252
    Mesh Size 100 Mesh
    Odor Odorless
    Quantity 150x150mm
    Chemical Name or Material Tantalum gauze, 100 mesh woven from 0.08mm (0.003 in.) dia. wire, twilled weave

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit