Alfa Aesar™ Tantalum nanopowder, APS 5-12nm, 99% (metals basis)

Manufacturer: Alfa Aesar™ 44994-03

Quantity 1g

 View more versions of this product

Catalog No. AA4499403

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD00011252
    UN Number UN1383
    Chemical Name or Material Tantalum nanopowder

    Specifications

    Health Hazard 2 GHS H Statement
    H228-H250-H252-H315-H319-H335
    Flammable solid.
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Causes skin irritation.
    Causes serious eye irritation.
    May
    TSCA TSCA
    Physical Form Powder
    UN Number UN1383
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P210-P222-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not allow contact with air.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove conta
    Quantity 1g
    Surface Area 41 to 51m2/g
    Particle Size APS 5 to 12nm
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC METALS, N.O.S.
    MDL Number MFCD00011252
    Chemical Name or Material Tantalum nanopowder
    Assay 99%, (metals basis)

    GHS H Statement
    H228-H250-H252-H315-H319-H335
    Flammable solid.
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Causes skin irritation.
    Causes serious eye irritation.
    May

    GHS P Statement
    P210-P222-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not allow contact with air.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove conta

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit