Tantalum powder, 99.97% (metal basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1034588

Quantity 2.5g

 View more versions of this product

Catalog No. AA1034588

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-25-7
    Physical Form Powder
    Assay Percent Range 99.97% (Metals basis, Nb 50ppm)
    MDL Number MFCD00011252
    Chemical Name or Material Tantalum

    Specifications

    Quantity 2.5g
    Color Gray to black
    DOT Information DOT Class: 4.1, Packaging Group: II
    EINECS Number 231-135-5
    Physical Form Powder
    Assay Percent Range 99.97% (Metals basis, Nb 50ppm)
    MDL Number MFCD00011252
    Appearance Gray to black powder
    CAS 7440-25-7
    Odor Odorless
    Health Hazard 1 H228-H315-H319-H335: Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Tantalum

    H228-H315-H319-H335: Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit