Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tantalum powder, 99.98% (metal basis), Alfa Aesar™, Puratronic™

Nb 50ppm

Manufacturer:  Alfa Aesar 1034488

Quantity 2.5g

 View more versions of this product

Catalog No. AA1034488

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7440-25-7
99.98% (Metals basis)
Tantalum
Powder
MFCD00011252

Specifications

2.5g
Gray to black powder
99.98% (Metals basis)
Odorless
Tantalum
7440-25-7
Powder
MFCD00011252
Gray to black

Safety and Handling

SDS & Safety Documents

H228-H315-H319-H335: Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-135-5

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit