Alfa Aesar™ Tantalum Thinfoil, 0.0025mm (0.0001 in.) thick, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 40716GH

Quantity 100x100mm

 View more versions of this product

Catalog No. AA40716GH

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-25-7
    MDL Number MFCD00011252
    Chemical Name or Material Tantalum Thinfoil, 0.0025mm (0.0001 in.) thick, Not certified pinhole free

    Specifications

    TSCA TSCA
    Physical Form Foil
    Weight ≈0.026g/25x25mm
    MDL Number MFCD00011252
    CAS 7440-25-7
    Quantity 100x100mm
    Odor Odorless
    Chemical Name or Material Tantalum Thinfoil, 0.0025mm (0.0001 in.) thick, Not certified pinhole free
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit