Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tantalum tris(diethylamido)-tert-butylimide, 99.99% (metals basis)

Precursor for TaN

Manufacturer:  Alfa Aesar™ 4341903

Quantity 1g

 View more versions of this product

Catalog No. AA4341903


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

169896-41-7
468.48
UN3129
Tantalum tris (diethylamido)-tert-butylimide
C16H39N4Ta
MFCD02684506
Tris(diethylamido)(tert-butylimido)tantalum(V)

Specifications

169896-41-7
C16H39N4Ta
MFCD02684506
Tris(diethylamido)(tert-butylimido)tantalum(V)
90°C (0.1mmHg)
1g
Tantalum tris (diethylamido)-tert-butylimide
Liquid
468.48
UN3129
Ampule
Yellow
Air & Moisture Sensitive
99.99%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE LIQUID, CORROSIVE, N.O.S.

storageNote1 :

  • Ampouled under argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit