Alfa Aesar™ Tantalum(V) bromide, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 1404109

Quantity 10g

 View more versions of this product

Catalog No. AA1404109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13451-11-1
    Formula Weight 580.49
    MDL Number MFCD00049563
    UN Number UN3260
    Synonym Tantalum pentabromide
    Chemical Name or Material Tantalum(V) bromide

    Specifications

    Formula Weight 580.49
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Pungent
    Physical Form Powder
    UN Number UN3260
    Quantity 10g
    TSCA TSCA
    Mesh Size −8 Mesh
    Sensitivity Moisture Sensitive
    MDL Number MFCD00049563
    CAS 13451-11-1
    Synonym Tantalum pentabromide
    Boiling Point 348.8°C
    Color Yellow
    Melting Point 265°C
    Health Hazard 1 Danger
    Density 4.67g/mL
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H312-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Chemical Name or Material Tantalum(V) bromide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H314-H318-H302-H312-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit