Tantalum(V) Chloride, 99.99% (Metal Basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1088014

Quantity 25g

 View more versions of this product

Catalog No. AA1088014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7721-01-9
    Physical Form Solid, Resublimed, Crystalline
    Assay Percent Range 99.99%, metal basis
    Molecular Formula TaCl5
    Formula Weight 358.21
    MDL Number MFCD00011253
    Chemical Name or Material Tantalum(V) Chloride, Ampouled under argon

    Specifications

    Formula Weight 358.21
    Quantity 25g
    DOT Information DOT Class: 8
    Health Hazard 2 When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
    EINECS Number 231-755-6
    Physical Form Solid, Resublimed, Crystalline
    Density 3.68
    Sensitivity Moisture sensitive
    MDL Number MFCD00011253
    CAS 7721-01-9
    Health Hazard 1 Harmful by inhalation, in contact with skin and if swallowed. Causes burns.
    Assay Percent Range 99.99%, metal basis
    Molecular Formula TaCl5
    Chemical Name or Material Tantalum(V) Chloride, Ampouled under argon

    When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

    Harmful by inhalation, in contact with skin and if swallowed. Causes burns.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit