Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tantalum(V) chloride, 99.99%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
7721-01-9
Cl5Ta
358.2
MFCD00011253
OEIMLTQPLAGXMX-UHFFFAOYSA-I
7721-01-9
Cl5Ta
358.2
MFCD00011253
OEIMLTQPLAGXMX-UHFFFAOYSA-I
For ultra trace analysis
Assay Percent Range 99.99%
Linear Formula TaCl5
UN Number 3260
Fieser 16,312; 17,321
Merck Index 15, 9186
Solubility Information Solubility in water: reacts with water. Other solubilities: soluble in alcohol,koh,h2so4
Formula Weight 358.2
Physical Form Crystals or Crystalline Powder
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: reacts with water. Other solubilities: soluble in alcohol, koh, h2so4
Total Trace Metal Impurities 100ppm max.
Boiling Point 242.0°C
Color White to Yellow
Melting Point 221.0°C
Quantity 5g
Chemical Name or Material Tantalum(V) chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 231-755-6