Alfa Aesar™ Tantalum(V) chloride, Puratronic™, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 1470806

Quantity 5g

 View more versions of this product

Catalog No. AA1470806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7721-01-9
    Formula Weight 358.21
    MDL Number MFCD00011253
    UN Number UN3260
    Merck Index 14,9054
    Chemical Name or Material Tantalum(V) chloride
    Grade Puratronic™

    Specifications

    Formula Weight 358.21
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Acidic
    Health Hazard 2 GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Sensitivity Moisture Sensitive
    Color White to pale yellow
    Melting Point 216°C
    Density 3.68g/mL
    Mesh Size −60 Mesh
    Physical Form Resublimed Powder
    MDL Number MFCD00011253
    UN Number UN3260
    Boiling Point 242°C
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    TSCA TSCA
    CAS 7721-01-9
    Merck Index 14,9054
    Quantity 5g
    Storage Note 1
    • Ampouled under argon
    RTECS Number WW5600000
    Chemical Name or Material Tantalum(V) chloride
    Assay 99.99%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit