Alfa Aesar™ Tantalum(V) ethoxide, 99.999% (metals basis), Nb <100ppm

Manufacturer: Alfa Aesar™ 1464309

Quantity 10g

 View more versions of this product

Catalog No. AA1464309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6074-84-6
    Linear Formula Ta(OCH2CH3)5
    Formula Weight 406.25
    MDL Number MFCD00049785
    UN Number UN2924
    Beilstein 3678999
    Chemical Name or Material Tantalum(V) ethoxide

    Specifications

    Formula Weight 406.25
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00049785
    CAS 6074-84-6
    Solubility Information Decomposes in water; Soluble in organic solvents
    Health Hazard 1 Danger
    Refractive Index 1.4880
    Flash Point 30° (87°F)
    Quantity 10g
    TSCA TSCA
    Density 1.566g/mL
    Physical Form Liquid
    Beilstein 3678999
    Boiling Point 145°C (0.1mmHg)
    Linear Formula Ta(OCH2CH3)5
    UN Number UN2924
    Health Hazard 2 GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Melting Point 18° to 19°C
    Concentration Nb <100ppm
    Sensitivity Air & Moisture Sensitive
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Chemical Name or Material Tantalum(V) ethoxide
    Assay 99.999%, (metals basis)

    GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit