Alfa Aesar™ Tantalum(V) isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol/hexane

Manufacturer: Alfa Aesar™ 40038AP

Quantity 500mL

 View more versions of this product

Catalog No. AA40038AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16761-83-4
    Linear Formula Ta[OCH(CH3)2]5
    Formula Weight 476.39
    MDL Number MFCD00145347
    UN Number UN1993
    Chemical Name or Material Tantalum(V) isopropoxide

    Specifications

    Formula Weight 476.39
    Health Hazard 2 GHS H Statement
    H225-H304-H361-H373-H315-H319-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P280h-P273-P305+P351+P338-P301+P330+P331-P315
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Get immediate medical advice/attention.
    Physical Form Liquid
    Linear Formula Ta[OCH(CH3)2]5
    Health Hazard 1 Danger
    UN Number UN1993
    Odor Petrol-like
    Quantity 500mL
    Sensitivity Moisture Sensitive
    MDL Number MFCD00145347
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    CAS 16761-83-4
    Concentration 10% w/v in isopropanol/hexane
    Chemical Name or Material Tantalum(V) isopropoxide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H225-H304-H361-H373-H315-H319-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P280h-P273-P305+P351+P338-P301+P330+P331-P315
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Get immediate medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit