Alfa Aesar™ Tantalum(V) methoxide

Manufacturer: Alfa Aesar™ 1883906

Quantity 5g

 View more versions of this product

Catalog No. AA1883906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 865-35-0
    Linear Formula Ta(OCH3)5
    Formula Weight 336.12
    MDL Number MFCD00058817
    UN Number UN3181
    Chemical Name or Material Tantalum(V) methoxide

    Specifications

    Formula Weight 336.12
    Health Hazard 3 GHS P Statement
    P210-P280h-P305+P351+P338
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Physical Form Crystalline
    Linear Formula Ta(OCH3)5
    MDL Number MFCD00058817
    Health Hazard 1 Warning
    Melting Point 50°C
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
    CAS 865-35-0
    UN Number UN3181
    Health Hazard 2 GHS H Statement
    H228-H302-H315-H319-H335
    Flammable solid.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Boiling Point 189°C (10mmHg)
    Quantity 5g
    Sensitivity Moisture Sensitive
    Chemical Name or Material Tantalum(V) methoxide

    GHS H Statement
    H228-H302-H315-H319-H335
    Flammable solid.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P280h-P305+P351+P338
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit