Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tantalum(V) methoxide

Manufacturer:  Alfa Aesar™ 1883914

Quantity 25g

 View more versions of this product

Catalog No. AA1883914


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

865-35-0
336.12
UN3181
Ta(OCH3)5
MFCD00058817
Tantalum(V) methoxide

Specifications

865-35-0
Ta(OCH3)5
MFCD00058817
189°C (10mmHg)
25g
Tantalum(V) methoxide
Crystalline
336.12
UN3181
50°C
Moisture Sensitive

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H302-H315-H319-H335
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P280h-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit