Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tantalum(V) oxide, 99.99%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
1314-61-0
O5Ta2
441.89
MFCD00011254
1314-61-0
O5Ta2
441.89
MFCD00011254
for ultra trace analysis
Assay Percent Range 99.99%
Linear Formula Ta2O5
Merck Index 15, 9188
Solubility Information Solubility in water: insoluble
Formula Weight 441.89
Physical Form Powder
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Total Trace Metal Impurities 0.01% max
Color Beige-White to White
Melting Point 1872°C
Quantity 10g
Chemical Name or Material Tantalum(V) oxide
Show More Show Less

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

EINECSNumber : 215-238-2