Alfa Aesar™ Tantalum(V) oxide, Puratronic™, 99.993% (metals basis excluding Nb), Nb 50ppm max

Manufacturer: Alfa Aesar™ 1088114

Quantity 25g

 View more versions of this product

Catalog No. AA1088114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-61-0
    Formula Weight 441.89
    MDL Number MFCD00011254
    Merck Index 14,9056
    Chemical Name or Material Tantalum(V) oxide
    Grade Puratronic™

    Specifications

    Formula Weight 441.89
    Physical Form Powder
    Melting Point 1800°C
    TSCA TSCA
    MDL Number MFCD00011254
    CAS 1314-61-0
    Merck Index 14,9056
    Quantity 25g
    Density 8.2g/mL
    RTECS Number WW5855000
    Odor Odorless
    Chemical Name or Material Tantalum(V) oxide
    Assay 99.993%, (metals basis excluding Nb), Nb 50ppm max.
    Grade Puratronic™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit