Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tantalum(V) tetraethoxide 2,4-pentanedionate, 99.99%, (metals basis), Thermo Scientific Chemicals

Catalog Number AA3963018
Click to view available options
Quantity:
2 g
10 g
50 g
20219-33-4
C13H27O6Ta
460.30
MFCD00145349
LSALXRIVOAYYJW-GLUPTGNJSA-M
tantalum v tetraethoxide 2,4-pentanedionate
122130355
[Ta+5].CC[O-].CC[O-].CC[O-].CC[O-].C\C([O-])=C\C(C)=O
20219-33-4
C13H27O6Ta
460.30
MFCD00145349
LSALXRIVOAYYJW-GLUPTGNJSA-M
tantalum v tetraethoxide 2,4-pentanedionate
122130355
[Ta+5].CC[O-].CC[O-].CC[O-].CC[O-].C\C([O-])=C\C(C)=O

Tantalum(V) tetraethoxide 2,4-pentanedionate is used in pharmaceutical, biochemical and chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 45°C
Density 1.5 g/mL
Boiling Point 95°C (0.5 mmHg)
Flash Point >65°C (149°F)
Quantity 50 g
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
IUPAC Name tantalum(5+) (2Z)-4-oxopent-2-en-2-olate tetraethanolate
Formula Weight 460.3
Percent Purity 99.99%
Physical Form Solid
Assay (metals basis)
Chemical Name or Material Tantalum(V) tetraethoxide 2,4-pentanedionate
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit