Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tantalum wire, 1.0mm (0.04 in.) dia., annealed, 99.95% (metals basis)

Click to view available options
Quantity:
1m
25m
5m
7440-25-7
Ta
180.948
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod
23956
CHEBI:33348
tantalum
[Ta]
7440-25-7
Ta
180.948
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum, elemental,hydride,tantalum-181,tantalum, metal and oxide dust,tantal,tantalum, metal,powder,foil,wire,rod
23956
CHEBI:33348
tantalum
[Ta]
Quantity 5m
Weight ≈13.0g/m
Odor Odorless
Physical Form Wire, ≈13.0g/m,Temper: soft
Percent Purity 99.95%
Chemical Name or Material Tantalum wire, 1.0mm (0.04 in.) dia., Annealed, Temper: soft
Assay (metals basis)

EINECSNumber : 231-135-5

TSCA : Yes

Recommended Storage : Ambient temperatures