Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tantalum wire, annealed, 99.9% (metals basis), Alfa Aesar™

Form: approx. 0.033g/m; Temper: soft

Manufacturer:  Alfa Aesar 40674G6

Quantity 10m

 View more versions of this product

Catalog No. AA40674G6


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-25-7
99.9% (Metals basis, Nb 50ppm)
Tantalum, Annealed
Wire, 0.05mm (0.002 in.) dia.
MFCD00011252

Specifications

10m
Gray to silver wire
99.9% (Metals basis, Nb 50ppm)
Odorless
Tantalum, Annealed
7440-25-7
Wire, 0.05mm (0.002 in.) dia.
MFCD00011252
Gray to silver

Safety and Handling

SDS & Safety Documents

EINECSNumber : 231-135-5

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit