Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TAOK1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA513851
Description
This antibody is predicted to react with mouse, rat and Xenopus based on sequence homology.
TAOK1 is a serine/threonine-protein kinase involved in regulation of the p38-containing stress-responsive MAP kinase pathway and extracellular signal-regulated protein kinase (ERK) kinases (MEKs). The activation of and binding to MEK3 by TAOK1 implicates TAOK1 in the regulation of the p38-containing stress-responsive MAP kinase pathway. A microtubule affinity-regulating kinase, TAOK1 (also known as MARKK) is an important regulator of mitotic progression, required for both chromosome congression and checkpoint-induced anaphase delay. TAOK1 is involved in various processes such as p38/MAPK14 stress-activated MAPK cascase, DNA damage response, and regulations of cytoskeleton stability. It phosphorylates MAP2K3, MAP2K6, and MARK2. TAOK1 acts as an activator of the p38/MAPK cascade by mediating phosphorylation and subsequent activation of the upsteam MAP2K3 and MAP2K6 kinases. It is involved in G-protein couples receptor signaling to p38/MAPK14. In response to DNA damage, TAOK1, is involved in the G2/M transition DNA damage checkpoint by activating the p38/MAPK14 stress-activated MAPK cascade by phosphorylaton of MAP2K3 and MAP2K6. In addition, it acts as a regulator of cytoskeleton stability and a regulator of apoptosis.
Specifications
TAOK1 | |
Polyclonal | |
Unconjugated | |
TAOK1 | |
2810468K05Rik; AU020252; D130018F14Rik; FLJ14314; h hTAOK1; H KIAA1361; hKFC-B; hTAOK1; KFC-B; Kiaa1361; Kinase from chicken homolog B; MAP3K16; MARK Kinase; Markk; microtubule affinity regulating kinase kinase; mKIAA1361; prostate-derived STE20-like kinase 2; prostate-derived sterile 20-like kinase 2; PSK2; PSK-2; serine/threonine protein kinase TAO1; serine/threonine protein kinase TAO1 homolog; serine/threonine-protein kinase TAO1; STE20-like kinase PSK2; tao; TAO kinase 1; TAO1; TAOK1; Thousand and one amino acid protein 1; thousand and one amino acid protein kinase 1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
57551 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q7L7X3 | |
TAOK1 | |
KLH conjugated synthetic peptide between 620-650 amino acids from the central region of human TAOK1. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction