Learn More
TAP-tag Rabbit, HRP, Polyclonal, GenScript™
Rabbit Polyclonal Antibody
Supplier: Genscript Corporation A0143540

Description
Reconstitute the lyophilized powder with deionized water (or equivalent) to an antibody concentration of 0.5 mg/mL. Well-characterized antibodies for epitope tags consisting of short sequences have been widely used in the study of protein expression in various systems.The TAP (tandem affinity purification) tag allows rapid purification of complexes from a relatively small number of cells without prior knowledge about the composition, activity, or function of the complex. Combined with mass spectrometry, the TAP strategy allows for the identification of proteins interacting with a given target protein. The Rabbit anti-TAP tag consists a 21 residue peptide, CSSGALDYDIPTTASENLYFQ, derived from c-terminal of the TAP construct after TEV cleavage.GenScript Rabbit Anti-TAP-tag [HRP] Polyclonal Antibody is highly purified from rabbit antiserum by immunoaffinity chromatography and then conjugated to horseradish peroxidase. The antibody is suitable for detecting fusion proteins that contain TAP tag.
TAP-tag Polyclonal antibody specifically detects TAP-tag in Tag samples. It is validated for ELISA, Western BlotSpecifications
TAP-tag | |
Polyclonal | |
PBS with 1% BSA and 0.01% thimerosal; pH 7.4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
-20°C or -80°C if preferred | |
Lyophilized | |
Tag |
ELISA, Western Blot | |
HRP | |
Tobacco Etch Virus | |
Synthetic peptide CSSGALDYDIPTTASENLYFQ conjugated to KLH. | |
40 μg | |
Primary | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.