Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAPA1 (CD81) Rat anti-Mouse, Clone: MT81, MilliporeSigma™

Rat Monoclonal Antibody

$153.40 - $387.00

Specifications

Antigen TAPA1 (CD81)
Clone MT81
Concentration 0.5 mg/mL
Applications Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Inhibition Assays, Neutralization, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS1947100
View Documents
MilliporeSigma
MABS1947100UL
100 μL
Each for $387.00
Add to cart
 
MABS194725
View Documents
MilliporeSigma
MABS194725UL
25 μL
Each for $153.40
Add to cart
 
Description

Description

TAPA1 (CD81) Monoclonal antibody specifically detects TAPA1 (CD81) in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Inhibition Assay, Neutralizing, Western Blotting.
Specifications

Specifications

TAPA1 (CD81)
0.5 mg/mL
Unconjugated
Rat
Mouse
Mouse L-M fibroblasts.
Primary
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
MT81
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Inhibition Assays, Neutralization, Western Blot
Purified
RUO
Purified rat monoclonal antibody IgG2b in PBS without azide.
Monoclonal
Protein G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit