Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte Tapwater Tour
Analyze the quality of tap water at home or in the classroom.
$71.75
Specifications
For Use With (Application) | Analyze the quality of tap water at home or in the classroom |
---|---|
Grade | 4 to 6 |
Sufficient For | 50 students |
Includes: Lecture materials, illustrated handouts, teacher tips, test procedures and wrap-up games
Catalog Number | Mfr. No. | Product Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Includes | Price | Quantity & Availability | ||||
S19119R
![]() |
LaMotte
R3608 |
Refill Kit | Lecture materials, illustrated handouts, teacher tips, test procedures and wrap up games |
Each for $71.75
|
|
S25705
![]() |
LaMotte
3608 |
Tap Water Tour Kit | Lecture materials, illustrated handouts, teacher tips, test procedures and wrap up games, five units |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Uses simple and safe reagent tablets in unit-dose form
- Includes five units: pH, chlorine, hardness, iron and copper
Order Info
Grades 4 to 6. Class size: 50 students.
Specifications
Analyze the quality of tap water at home or in the classroom | |
4 to 6 | |
50 students |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title