Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Taq CORE Kit 25

Supplied with standard reaction buffers

Manufacturer:  MP Biomedicals™ 11EPTQK25L

 View more versions of this product

Catalog No. 11EPTQK25L


Add to cart

Includes: Taq DNA Polymerase 5U/µL, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 25mM dNTP Mix

Description

Description

Specifications

Specifications

5 x 1000U
Store at -20°C
Taq DNA Polymerase 5U/μL, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 25mM dNTP Mix
PCR
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit