Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Taq-&GO Ready-to-Use PCR Mix

A ready-to-use, optimized 5X reaction mix for PCR

Manufacturer:  MP Biomedicals™ 11EPTAG110

 View more versions of this product

Catalog No. 11EPTAG110


Add to cart

Description

Description

Simple-to-use: add 10µL of Taq-&GO to a 40µL volume containing desired template and primers and reaction mix is ready.
Unit Definition: One unit incorporates 10nmol of total dNTPs into acid-precipitable DNA in 30 min. at 74°C.

Specifications

Specifications

10 x 1mL
Store at -20°C.
Standard PCR
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit