Learn More
TARDBP Recombinant Protein, Abnova
Human TARDBP full-length ORF (AAH01487.1, 1 a.a. - 260 a.a.) recombinant protein with GST-tag at N-terminal
Manufacturer:  Abnova Corporation H00023435-P01-25
 View more versions of this product
Catalog No. 89-012-331
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Antibody Production, ELISA, Protein Array, Western Blotting (Recombinant Protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Human TARDBP Full-length ORF Recombinant Protein with GST-tag at N-terminal
In vitro wheat germ expression system
12.5% SDS-PAGE stained with Coomassie Blue
Wheat Germ (in vitro)
Human
Solution
54.34
8
Glutathione Sepharose 4 Fast Flow
25μg
-80°C
Yes