Learn More
3M™ Tartan™ Box Sealing Tape
Tartan™ Box Sealing Tape is a general purpose packing tape ideal for non-critical box sealing applications
Manufacturer:  3M™ 00021200615535
Catalog No. 19-072-022
Add to cart
Description
Description
  • General purpose packaging tape for light-duty applications on a wide range of materials
  • High edge tear and split resistance ideal for general purpose applications
  • Polypropylene backing with synthetic rubber adhesive delivers conformability under stress
  • Film backing is easy to unwind and resists abrasion, moisture and scuffing.
  • Backing easily conforms around edges and on rough surfaces for a tight seal.
  • Easy unwind and smooth dispensing works well on high-speed automated packaging lines
  • Excellent adhesion to recycled liner paper and fiberboard
Specifications
Specifications
Clear
48 mm x 100 m
Hot Melt Synthetic Rubber Resin
Hot Melt Synthetic Rubber Resin
Box Sealing Tape
Automated Carton Sealing, Box Closure, Cold Storage and Tamper Evident
36/case