Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Lab Vision™ Tartrate Resistant Acid Phosphatase (TRAP) Ab-1, Mouse Monoclonal Antibody
Tartrate Resistant Acid Phosphatase (TRAP) Ab-1, Mouse MAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Supplier: Epredia™ MS1768R1
Description
Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher's disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin's lymphoma and in the sera of patients undergoing active bone turnover.Host Species: Mouse
Clone: 26E5
Isotype: IgG2b
Species Reactivity: Human. Others not known.
Epitope: N-terminal
Immunogen: Recombinant protein corresponding to N-terminal region of human TRAP
Positive Control: Lung (macrophages)
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Tartrate Resistant Acid Phosphatase (TRAP) Ab-1 | |
Monoclonal | |
Unconjugated | |
Recombinant protein corresponding to N-terminal region of human TRAP | |
IVD | |
Primary | |
Prediluted |
Immunohistochemistry (Paraffin) | |
26E5 | |
Mouse | |
1 mL | |
Cancer and Tumor Biology | |
Human | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction