Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAS 301, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
193620-69-8
C23H19NO3
357.409
KUEYYIJXBRWZIB-UHFFFAOYSA-N
3-bis 4-methoxyphenyl methylene-2-indolinone,3-bis-4-methoxphenyl methylene-2-indolinone,tas hplc , solid,3-bis 4-methoxyphenyl methylene-oxindole,3-bis 4-methoxyphenyl methylidene-1h-indol-2-one,3-bis-4-methoxy-phenyl-methylene-1,3-dihydro-indol-2-one,3-bis 4-methoxyphenyl methylidene-2,3-dihydro-1h-indol-2-one,3r-3-benzyl-n,n',n'-trimethyl-3-piperidinecarbohydrazide hydrochloride
9885087
3-[bis(4-methoxyphenyl)methylidene]-1H-indol-2-one
COC1=CC=C(C=C1)C(=C2C3=CC=CC=C3NC2=O)C4=CC=C(C=C4)OC
This item is not returnable. View return policy
193620-69-8
C23H19NO3
357.409
KUEYYIJXBRWZIB-UHFFFAOYSA-N
3-bis 4-methoxyphenyl methylene-2-indolinone,3-bis-4-methoxphenyl methylene-2-indolinone,tas hplc , solid,3-bis 4-methoxyphenyl methylene-oxindole,3-bis 4-methoxyphenyl methylidene-1h-indol-2-one,3-bis-4-methoxy-phenyl-methylene-1,3-dihydro-indol-2-one,3-bis 4-methoxyphenyl methylidene-2,3-dihydro-1h-indol-2-one,3r-3-benzyl-n,n',n'-trimethyl-3-piperidinecarbohydrazide hydrochloride
9885087
3-[bis(4-methoxyphenyl)methylidene]-1H-indol-2-one
COC1=CC=C(C=C1)C(=C2C3=CC=CC=C3NC2=O)C4=CC=C(C=C4)OC

Inhibitor of smooth muscle cell migration and proliferation

Inhibitor of smooth muscle cell migration and proliferation. Blocks voltage-independent calcium influx and downstream PKC signaling. Inhibits neointimal thickening after balloon catheter injury to the rat common carotid artery.
Quantity 10mg
Formula Weight 357.4
Percent Purity >99%
Chemical Name or Material TAS 301

Recommended Storage : Store at +4°C