Learn More
Novus Biologicals™ TAS2R39 Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP233684PEP
Catalog No. NBP23684PEP
Add to cart
Description
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human TAS2R39. The TAS2R39 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
Antibody Competition
259285
>80% by SDS-PAGE and Coomassie blue staining
E. coli
RUO
TAS2R39
PBS and 1M Urea, pH 7.4.
21 kDa
100 μl
Store at -20°C. Avoid freeze-thaw cycles.
TAS2R39
Human