Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAS2R4 Polyclonal Antibody
TAS2R4 Polyclonal Antibody
Supplier: Thermo Scientific PA567752
Description
TAS2R4 Polyclonal Antibody for Western Blot

Specifications
TAS2R4 | |
Polyclonal | |
Unconjugated | |
TAS2R4 | |
bitter taste receptor T2R16; candidate taste receptor T2R4; candidate taste receptor T2R8; MGC163311; mGR08; mt2r8; T2R108; T2R16; T2R20; T2R4; T2R8; Tas2r108; TAS2R4; tas2r4 {ECO:0000250; Tas2r8; taste 2 receptor member 4; taste receptor T2R8; taste receptor type 2 member 20; Taste receptor type 2 member 4; taste receptor type 2 member 8; taste receptor, type 2, member 108; taste receptor, type 2, member 4; UniProtKB:Q9JKT3} | |
Rabbit | |
affinity chromatography | |
RUO | |
50832, 554302, 57253 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q67ET0, Q9JKT3, Q9NYW5 | |
Tas2r108, TAS2R4 | |
Synthesized peptide derived from human TAS2R4. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction