Learn More
Tocris Bioscience™ TASP 0390325 Available on GSA/VA Contract for Federal Government customers only.
Potent and selective V1B receptor antagonist; orally bioavailable
$215.50 - $901.50
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
57-291-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5729/10
Each for $215.50
Add to cart
57-295-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5729/50
Each for $901.50
Add to cart
Description
Description
Potent and selective V1B receptor antagonist (IC50 = 2.22 nM for rat pituitary receptor). Exhibits no significant affinity for 85 other receptors, ion channels and transporters at 10 μM, including V1A and V2. Increases ACTH levels in rats. Orally bioavailable. Antidepressant and anxiolytic.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.