Learn More
Aviva Systems Biology Corp TASP1 Antibody - middle region (ARP57018_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP57018_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TASP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TASP1. Alternate names: C20orf13, FLJ20212, MGC39159, dJ585I14.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62061