Learn More
TASP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 167391AP
Catalog No. 50-173-1149
Add to cart
Description
Description
This gene encodes an endopeptidase that cleaves specific substrates following aspartate residues. The encoded protein undergoes posttranslational autoproteolytic processing to generate alpha and beta subunits, which reassemble into the active alpha2-beta2 heterotetramer. It is required to cleave MLL, a protein required for the maintece of HOX gene expression, and TFIIA, a basal transcription factor. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications
Specifications
TASP1
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8R1G1, Q9H6P5
C20orf13, TASP1, Taspase 1, Threonine aspartase 1
Rabbit
IgG
150 μL
-20°C
Primary
311468, 55617, 75812
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TASP1
Liquid
Tasp1
TASP1 Fusion Protein Ag9918
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat