Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TASP1 Mouse anti-Human, Clone: OTI1H6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525378

Catalog No. PIMA525378


Add to cart

Description

Description

This gene encodes an endopeptidase that cleaves specific substrates following aspartate residues. The encoded protein undergoes posttranslational autoproteolytic processing to generate alpha and beta subunits, which reassemble into the active alpha2-beta2 heterotetramer. It is required to cleave MLL, a protein required for the maintece of HOX gene expression, and TFIIA, a basal transcription factor. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications

Specifications

TASP1
Monoclonal
0.54 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H6P5
Taspase, threonine aspartase, 1, C20orf13, FLJ20212, MGC39159, dJ585I14.2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55617
Western Blot
OTI1H6
Unconjugated
TASP1
Liquid
TASP1
Full length human recombit protein of TASP1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit