Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TAT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA526541
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
TAT is a mitochondrial protein tyrosine aminotransferase which is present in the liver and catalyzes the conversion of L-tyrosine into p-hydroxyphenylpyruvate.
Specifications
TAT | |
Polyclonal | |
Unconjugated | |
TAT | |
2.6.1.5; L-tyrosine:2-oxoglutarate aminotransferase; Tat; testis tissue sperm-binding protein Li 34a; tyrosine aminotransferase; tyrosine aminotransferase, cytosolic; tyrosine transaminase | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
234724, 6898 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P17735, Q8QZR1 | |
TAT | |
KLH conjugated synthetic peptide between 279-306 amino acids from the central region of human TAT. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction