Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TATA Binding Protein Rabbit anti-Human,Mouse,Rat, Unconjugated, Clone: 5J13, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen TATA Binding Protein
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB110625MI
View Documents
MilliporeSigma
ZRB110625UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB11064XMI
View Documents
MilliporeSigma
ZRB11064X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

TATA Binding Protein Monoclonal antibody specifically detects TATA Binding Protein in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

TATA Binding Protein
Unconjugated
Rabbit
TATA sequence-binding protein;TATA-binding factor;TATA-box factor;Transcription initiation factor TFIID TBP subunit
GST/Hig-tagged recombinant fragment corresponding to the first 57 amino acids from the N-terminal region of human TATA-box-binding protein.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse, Rat
TBP, GTF2D1, TF2D, TFIID
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit