Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau Ab-2 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Tau Ab-2, Mouse Monoclonal Antibody.

$632.00 - $1160.00

Specifications

Antigen TAU Ab-2
Clone TAU-5
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Form Concentrated
View More Specs

Products 7
Catalog Number Mfr. No. Conjugate Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Conjugate Concentration Quantity Price Quantity & Availability  
MS247B1
View Documents
Epredia
MS247B1
Biotin 200μg/mL 500 μL
Each for $839.00
Add to Cart
 
MS247P1
View Documents
Epredia
MS247P1
Unconjugated 200μg/mL 500 μL
Each for $632.00
Add to Cart
 
MS247P
View Documents
Epredia
MS247P
Unconjugated 200μg/mL 1 mL
Each for $1,160.00
Add to Cart
 
MS247P1ABX
View Documents
Epredia
MS247P1ABX
Unconjugated 1mg/mL 100 μL
Each for $870.00
Add to Cart
 
MS247PABX
View Documents
Epredia
MS247PABX
Unconjugated 1mg/mL 200 μL
Each for $1,044.00
Add to Cart
 
MS247B0
View Documents
Epredia
MS247B0
Biotin 200μg/mL 100 μL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
MS247B
View Documents
Epredia
MS247B
Biotin 200μg/mL 1 mL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

Tau proteins are microtubule-associated proteins (MAPs). Alternate splicing of tau mRNA and differential phosphorylation contribute to the heterogeneity of tau.

Host Species: Mouse

Clone: TAU-5

Isotype: IgG1

Species Reactivity: Human, Sheep, Cow, Mouse and Rat. Others not known.

Epitope: Middle of Tau

Immunogen: Purified bovine microtubule-associated proteins

Molecular Weight: 45-68kDa

Positive Control: IMR5 cells, Human T98G glioblastoma cells or Alzheimer's disease brain tissue

Cellular Localization: Cytoplasmic Ab-2 reacts with the non-phosphorylated as well as the phosphorylated forms of tau and its epitope maps in the middle of tau. Ab-2 intensely stains the human neurofibrillary tangles, neuropil threads, and neuritic plaques associated with Alzheimer's disease. Ab-2 shows no cross reaction with other MAPs or tubulin.

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

TAU Ab-2
Immunofluorescence, Immunoprecipitation, Western Blot
Concentrated
RUO
Bovine, Human, Mouse, Rat, Sheep
IgG1
TAU-5
Monoclonal
Mouse
Neuroscience
Purified bovine microtubule-associated proteins
Primary
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit