Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tau Rabbit anti-Human, Clone: SN62-09, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267702
Description
Tau Monoclonal antibody specifically detects Tau in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Tau | |
SN62-09 | |
Unconjugated | |
Purified | |
DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau | |
MAPT | |
Synthetic phospho-peptide surrounding S396 of Tau | |
Monoclonal | |
Protein A purified | |
Alzheimers Research, Apoptosis, Chaperone Mediated Autophagy (CMA), Cytokine Research, Cytoskeleton Markers, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers | |
Human,Mouse,Rat |
Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1:500-1:1000 Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
TBS (pH7.4), 1%BSA, 40% Glycerol with 0.05% Sodium Azide | |
4137 | |
Rabbit | |
IgG | |
Primary | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title