Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau tubulin kinase 2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Tau tubulin kinase 2
Applications Immunofluorescence
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095437
View Documents
Novus Biologicals
NBP303311100UL
100 μg
Each for $320.71
Add to cart
 
NB095438
View Documents
Novus Biologicals
NBP30331120UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

Tau tubulin kinase 2 Polyclonal antibody specifically detects Tau tubulin kinase 2 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

Tau tubulin kinase 2
Unconjugated
Rabbit
Protein Kinase
PBS with 50% glycerol, pH7.3.
146057
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence
Purified
RUO
Human
EC 2.7.11, EC 2.7.11.1, KIAA0847tau-tubulin kinase 2, SCA11, spinocerebellar ataxia 11, tau tubulin kinase 2, TTBK
Recombinant fusion protein containing a sequence corresponding to amino acids 1185-1244 of human Tau tubulin kinase 2 (NP_775771.3).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit