Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Taurine, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
100 g
1 kg
5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
107-35-7
C2H7NO3S
125.14
XOAAWQZATWQOTB-UHFFFAOYSA-N
2-aminoethane-1-sulfonic acid
NCCS(O)(=O)=O
107-35-7
C2H7NO3S
125.14
XOAAWQZATWQOTB-UHFFFAOYSA-N
2-aminoethane-1-sulfonic acid
NCCS(O)(=O)=O

T2860, 107-35-7, C2H7NO3S

Spectrum™ Taurine, USP is an organic acid found in animal tissues, specifically bile and the large intestine. Some of its roles include conjugation of bile acids, membrane stabilization and modulation of calcium signaling. It is crucial for cardiovascular function. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Bottle
Ignition Residue 0.003
Quantity 100 g
Loss on Drying 0.003
CAS Max % 1
Percent Purity 98 to 102%
Grade USP